Learn More
Hanna Instruments™ Ozone Reagent Water Testing Kit
Product Code. 11867930
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
Product Code. 11867930
Brand: Hanna Instruments™ HI9375701
Only null left
Add to basket
Add to basket
Specifications
No. of Tests 100
Quantity 100/Pk.
Range 0 to 2mg/L
Test Method DPD
Type Ozone Reagent
compliance-icons compliance-icons
Product Identifier
  • X100 REAGENT OZONE
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide, EDTA