Learn More
Hanna Instruments™ Ozone Reagent Water Testing Kit
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
Specifications
Specifications
| No. of Tests | 100 |
| Quantity | 100/Pk. |
| Range | 0 to 2mg/L |
| Test Method | DPD |
| Type | Ozone Reagent |
Safety and Handling
Product Identifier
- X100 REAGENT OZONE
- Danger
- Acute toxicity Category 4
- Specific target organ toxicity Category 1
- H332-Harmful if inhaled.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : potassium iodide, EDTA