missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxymetazoline-d4 (imidazoline-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Oxymetazoline dihydroimidazolyl-d4 |
| CAS | 1262053-60-0 |
| Molecular Formula | C16 2H4 H20 N2 O |
| Quantity | 0.005 g |
| Synonym | Phenol, 3-[(4,5-dihydro-4,5-d2-1 H-imidazol-2-yl-4,5-d2)methyl]-6-(1,1-dimethylethyl)-2,4-dimethyl-, 3-[(4,5-Dihydro-4,5-d2-1 H-imidazol-2-yl-4,5-d2)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol, Oxymetazoline-d4, Oxymetazoline-D4 (dihydroimidazolyl D4) |
| InChI Formula | InChI=1 S/C16H24N2O/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14/h8,19 H,6-7,9H2,1-5H3,(H,17,18)/i6D2,7D2 |
| SMILES | [2 H]C1([2 H])NC(=NC1([2 H])[2 H])Cc2c(C)cc(c(O)c2C)C(C)(C)C |
| IUPAC Name | 6-tert-butyl-2,4-dimethyl-3-[(4,4,5,5-tetradeuterio-1 H-imidazol-2-yl)methyl]phenol |
| Molecular Weight (g/mol) | 264.4 |
| Formula Weight | 264.214 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction