Learn More
Chemetrics™ Oxygen ULR CHEMets™ Refill
Product Code. 11737073
Change view
Click to view available options
Quantity:
30 Ampoules
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11737073 30 Ampoules Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11737073 Supplier Chemetrics™ Supplier No. R7511
 Restricted Item
Estimated Shipment: 23-11-2026
Specifications
Analytes Oxygen, dissolved
For Use With (Application) Dissolved oxygen analysis
For Use With (Equipment) Refill for kit K-7511; compatible with C-7511 comparators
Includes 30 self-filling ULR CHEMets ampoules
No. of Tests 30
Quantity 30 Ampoules
compliance-icons compliance-icons
Product Identifier
  • Oxygen, dissolved
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician/first aider.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to authorised chemicallandfill or if organic to high temperature incineration.