Learn More
Oxybutynin Chloride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Oxybutynin Hydrochloride |
| CAS | 1508-65-2 |
| Molecular Formula | C22 H31 N O3 . Cl H |
| Quantity | 1 g |
| Synonym | Oxybutynin Hydrochloride, Benzeneacetic acid','α-cyclohexyl-α-hydroxy-','4-(diethylamino)-2-butynyl ester','hydrochloride (9CI), Cyclohexaneglycolic acid','α-phenyl-','4-(diethylamino)-2-butynyl ester','hydrochloride (7CI,8CI), 2-Butyn-1-ol','4-(diethylamino)-','α-phenylcyclohexaneglycolate (ester)','hydrochloride (8CI), 4-(Diethylamino)-2-butynyl α-phenylcyclohexaneglycolate hydrochloride, Cystrin, Dridase, MJ 4309-1, MJ 50-58, Oxybutynin chloride, Oxybutynin hydrochloride, Pollakisu, Tropax |
| InChI Formula | InChI=1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H |
| SMILES | Cl.CCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2 |
| IUPAC Name | 4-(diethylamino)but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride |
| Molecular Weight (g/mol) | 393.95 |
| Formula Weight | 393.2071 |