Learn More
Thermo Scientific™ Oxoid™ VET-RPLA Toxin Detection Kit
Click to view available options
Quantity:
20 Tests/Kit
Unit Size:
20 tests
Description
Thermo Scientific™ Oxoid VET-RPLA Toxin Detection Kit uses reverse passive latex agglutination to detect E. coli heat-labile enterotoxin and Vibrio cholerae enterotoxin.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | VET-RPLA Toxin Detection Kit |
| Detectable Analytes | E. coli enterotoxin, Vibrio cholerae enterotoxin |
| Quantity | 20 Tests/Kit |