Learn More
Thermo Scientific™ Oxoid™ SPS Disc
Product Code. 10127462
Change view
Click to view available options
Quantity:
50 Disks/Cartridge
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10127462 50 Disks/Cartridge Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10127462 Supplier Thermo Scientific™ Supplier No. DD0016T
  • ONLINE EXCLUSIVE PRICE
    40.95€
    12.59€ / Pack of 50
    Save 28.36€
    69% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Susceptibility disc format
TRUSTED_SUSTAINABILITY
Specifications
Description SPS Disk
Detectable Analytes P. anaerobius
Format Susceptibility Disk
Type SPS Disk
Quantity 50 Disks/Cartridge