Learn More
Thermo Scientific™ Oxoid™ PBP2' Latex Agglutination Test Kit
Product Code. 10699164
Change view
Click to view available options
Quantity:
50 Tests/Kit
Unit Size:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10699164 50 Tests/Kit 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10699164 Supplier Thermo Scientific™ Supplier No. DR0900A
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Edge
Add to basket
For detection of PBP2' and identification of methicillin resistance in Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Thermo Scientific™ Oxoid PBP2' Latex Agglutination Test is a rapid latex slide agglutination test for detection of PBP2' as an aid in identifying methicillin resistance in both Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Ordering Alerts:
50 tests/kit.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Latex Agglutination Test Kit
For Use With (Application) Healthcare
Quantity 50 Tests/Kit