Learn More
Thermo Scientific™ Biochemical Identification System (O.B.I.S.) Campy
Beskrivelse
Rapidly differentiate Campylobacter, Helicobacter, and Acrobacter species from other Gram-negative organisms with this biochemical identification system.
Rapidly differentiate Campylobacter, Helicobacter and Acrobacter species from other Gram-negative organisms with Thermo Scientific™ Oxoid™ Biochemical Identification System (O.B.I.S.) campy. This rapid colorimetric test delivers simple and rapid results for the detection of L-alanyl aminopeptidase in Gram-negative organisms. It has been designed for the differentiation of Campylobacter, Helicobacter, and Arcobacter species from other Gram-negative organisms and incorporates a Gram-lysis test which will demonstrate Gram status.
Use O.B.I.S. campy for rapid differentiation of species of Campylobacter, Helicobacter, and Arcobacter from all other Gram-negative organisms1.
- Simple—easy to implement
- Rapid—results in 6 minutes of testing suspect colonies
The O.B.I.S. campy test differentiates species of Campylobacter, Helicobacter, and Arcobacter from all other Gram-negative organisms1. Unlike other Gram-negative organisms, Campylobacteraceae do not possess the enzyme L-alanyl aminopeptidase (L-ALA). The O.B.I.S. campy test incorporates a rapid test to detect this enzyme and a Gram-lysis reagent to rapidly determine the Gram status. First, the Gram-lysis test (or a Gram stain) must be carried out. This test differentiates between Gram-positive and Gram-negative bacteria2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Tekniske data
Tekniske data
| Description | O.B.I.S. campy |
| Detectable Analytes | Campylobacter Antigen |
| Quantity | 60 Tests/Kit |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.