Learn More
Thermo Scientific™ Oxoid™ Nitrocefin Solution
Product Code. 10280333
Change view
Click to view available options
Quantity:
10 Vial Set/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10280333 10 Vial Set/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10280333 Supplier Thermo Scientific™ Supplier No. SR0112C
  • ONLINE EXCLUSIVE PRICE
    1635.00€
    869.59€ / Pack of 10
    Save 765.41€
    46% Off
 Restricted Item
Estimated Shipment: 03-12-2026
The Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
  • Reconstitutes to 500μg/mL
  • Contents: 5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL)
TRUSTED_SUSTAINABILITY
Specifications
Description Nitrocefin
Format Vial
Includes 5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL)
Packaging Type 2 x 5 Vials
Quantity 10 Vial Set/Pk.