Learn More
Thermo Scientific™ EC Broth with MUG (Dehydrated)
Product Code. 13265909
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13265909 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13265909 Supplier Thermo Scientific™ Supplier No. CM0979B
Selectively detect Escherichia coli from food and environmental samples with this selective medium.
Selectively detect Escherichia coli from food and environmental samples with Thermo Scientific™ Oxoid™ EC Broth with MUG (Dehydrated). EC Broth is a selective medium for the differentiation of fecal coliforms and the confirmatory test for Escherichia coli from food and environmental samples1,2. It is also suitable for the enumeration of presumptive E. coli in milk and milk products using the most probable number technique3. The MUG (4-methylumbelliferyl-ß-D-glucuronide) is cleaved by the enzyme glucuronidase (GUD) produced by Escherichia coli resulting in a fluorescent end-product.
Escherichia coli is a common Gram-negative microorganism which may be present in food and water samples. The presence of E.coli cells may be indicative of fecal contamination, although their ingestion does not necessarily have adverse effects on health. Nevertheless, certain strains are responsible for diarrhoeal disease and can also lead to more serious forms of illness.
EC Broth has shown to be a reliable medium producing dependable results in 24 to 48 hours3.
  • Selective: the addition of bile salts no.3 in the medium is inhibitory to gram-positive bacteria, especially bacilli and fecal Streptococci making the medium selective for Escherichia coli
  • Easy-to-read: gas production as a result of lactose fermentation is a presumptive evidence of the presence of coliform bacteria.
  • Recommended by FDA/BAM: for the confirmation of Escherichia coli from shellfish and meat4.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial reference: FDA.
Description EC Broth w/MUG
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 13.5L medium
For Laboratory Use Only