Learn More
Thermo Scientific™ Oxoid™ Campylobacter Selective Agar (Preston)
Description
Specifically designed for the selective isolation of Campylobacter species, this high-performance agar ensures accuracy and reliability in your results.
Specifications
Specifications
| Color | Black |
| Content And Storage | 2-12°C |
| Format | 90 mm |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Volume | 17 mL |
| Fill Weight | 17mL |
| Packaging Type | 1 x 10 plates |
| Quantity | 10 plates |