missing translation for 'onlineSavingsMsg'
Learn More

Oxociprofloxacin, TRC

Product Code. 30409916
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409916 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409916 Supplier Toronto Research Chemicals Supplier No. TRCO84950010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxociprofloxacin
CAS 103237-52-1
Purity Grade Notes HPLC
Molecular Formula C17 H16 F N3 O4
Quantity 10 mg
Synonym 3-Quinolinecarboxylic acid','1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(3-oxo-1-piperazinyl)-, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(3-oxo-1-piperazinyl)-3-quinolinecarboxylic acid, 3'-Oxociprofloxacin, 4-Oxociprofloxacin, BAY-q 3542, Oxociprofloxacin
InChI Formula InChI=1S/C17H16FN3O4/c18-12-5-10-13(6-14(12)20-4-3-19-15(22)8-20)21(9-1-2-9)7-11(16(10)23)17(24)25/h5-7,9H,1-4,8H2,(H,19,22)(H,24,25)
SMILES OC(=O)C1=CN(C2CC2)c3cc(N4CCNC(=O)C4)c(F)cc3C1=O
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-(3-oxopiperazin-1-yl)quinoline-3-carboxylic acid
Molecular Weight (g/mol) 345.325
Formula Weight 345.1125
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.