Learn More
Diversey™ Oxivir Excel Wipes CE
Description
- One-step cleaner disinfectant with broad spectrum efficacy and short contact times
- Oxivir Excel Wipe can be used on non-invasive medical devices
- Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 seconds
- Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
- No hazard classification
- AHP formulation is gentle on surfaces extending equipment life
- Excellent user safety profile - no PPE required
- Does not contain respiratory irritants
- Suitable for use in high risk areas including ICU, OR, NICU and patient rooms
- Fragrance free
- No rinsing required
- Safer for the environment and user, since the AHP actives break down into water and oxygen. Free from Volatile Organic Compounds (VOCs)
- Medical Device Class IIa
Specifications
Specifications
| Product Type | All Purpose Wipes |
| Length (English) | 6 in. |
| Length (Metric) | 15 cm |
| Width (English) | 7 in. |
| Width (Metric) | 17 cm |
| Material | Polypropylene |
| Description | For BE, ES, IT, NL, and PT only |
| Dimensions (L x W) | 15 x 17 cm (6 x 7 in.) |
| Format | Canister, PT02 |
| For Use With (Application) | All purpose virucidal wipes |
| Show More |
The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004