missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ Oxivir Excel Wipes
Click to view available options
Dimensions (L x W):
15 x 17 cm
27 x 20 cm
Format:
Canister, PT02
Flow Pack
Packaging Quantity:
100 Wipes/Pk., 12 Pk.
160 Wipes/Pk., 6 Pk.
Unit Size:
Pack of 1200
Pack of 960
Description
- One-step cleaner disinfectant with broad spectrum efficacy and short contact times
- Extensively tested against European norms and effective against multiple pathogens of concern (Bacteria, Viruses and Yeasts)
- Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 sec
- Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
- No hazard classification
- Powered by AHP Technology (Accelerated Hydrogen Peroxide)
- AHP formulation is gentle on surfaces extending equipment life
- Excellent user safety profile - no PPE required
- Does not contain respiratory irritants
- Suitable for use in high risk areas including ICU, OR, NICU and patient rooms
- Fragrance free
- No rinsing required
- Safer for the environment and user, since the AHP actives break down into water and oxygen. Free from Volatile Organic Compounds (VOCs)
Specifications
Specifications
| Product Type | All Purpose Wipes |
| Length (English) | 6 in. |
| Length (Metric) | 15 cm |
| Width (English) | 7 in. |
| Width (Metric) | 17 cm |
| Material | Polypropylene |
| Description | For BE and PT only |
| Dimensions (L x W) | 15 x 17 cm |
| Format | Canister, PT02 |
| For Use With (Application) | All purpose virucidal wipes |
| Show More |
The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction