Learn More
Diversey™ Oxivir Excel Wipes
Click to view available options
Dimensions (L x W):
15 x 17 cm
27 x 20 cm
Format:
Canister, PT02
Flow Pack
Packaging Quantity:
100 Wipes/Pk., 12 Pk.
160 Wipes/Pk., 6 Pk.
Unit Size:
Pack of 1200
Pack of 960
Description
- One-step cleaner disinfectant with broad spectrum efficacy and short contact times
- Extensively tested against European norms and effective against multiple pathogens of concern (Bacteria, Viruses and Yeasts)
- Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 sec
- Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
- No hazard classification
- Powered by AHP Technology (Accelerated Hydrogen Peroxide)
- AHP formulation is gentle on surfaces extending equipment life
- Excellent user safety profile - no PPE required
- Does not contain respiratory irritants
- Suitable for use in high risk areas including ICU, OR, NICU and patient rooms
- Fragrance free
- No rinsing required
- Safer for the environment and user, since the AHP actives break down into water and oxygen. Free from Volatile Organic Compounds (VOCs)
Specifications
Specifications
| Product Type | All Purpose Wipes |
| Length (English) | 10 in. |
| Length (Metric) | 26 cm |
| Width (English) | 8 in. |
| Width (Metric) | 20 cm |
| Material | Polypropylene |
| Description | For BE, IT , PT only |
| Dimensions (L x W) | 27 x 20 cm |
| Format | Flow Pack |
| For Use With (Application) | All purpose virucidal wipes |
| Show More |
The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004