Learn More
WTW™ OxiTop™ IS 12 Measuring Head
Product Code. 10623742
Click to view available options
Weight (Metric):
11 kg
Unit Size:
Each
Product Code. 10623742
Brand: WTW™ 208211
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Respirometric OxiTop™ measuring system for BOD self check measurement with undiluted samples – measuring ranges up to 4,000 mg/L BOD possible
  • Easy handling without dilution series
  • Data security by built-in measured value memory
  • Suitabe for immediate use
TRUSTED_SUSTAINABILITY
Specifications
Includes BOD bottles with amber, magnetic bars, bar puller, sodium hydroxide pellets, NaOH tanks, volumetric flasks 164 and 432mL.
For Use With (Application) Measurement using OxiTop™ is based on pressure measurement in a closed system
Certifications/Compliance CE
Weight (Metric) 11 kg
Warranty 1 Year
Temperature (Metric) Operating 5°C to 50°C
Length (Metric) 42 cm
Height (Metric) 28 cm
Accuracy (DO) ±1%
Electrical Requirements 100/240V, 50/60Hz
Voltage 100/240 V
Frequency 50/60 Hz