Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
Description
- OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
- Measuring ranges up to 4000 mg/L BOD possible
- No need to convert data - direct input of sample volume
- Direct display of the BOD value on the measuring head screen
- Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
Specifications
Specifications
| Type | OxiTop™-i IS 12 |
| Display Type | Backlit Graphic LCD |
| Range | Measuring ranges up to 4000 mg/L BOD possible |
| Includes | Twelve OxiTop™-i measuring heads, universal plug-in power supply 100-240 VAC, 50/60 Hz |