Learn More
WTW™ OxiTop™ IS BOD Self Check Measurement System
Product Code. 11751243
Click to view available options
Type:
BOD Meter, 2 Heads
BOD Meter, 6 Heads
Unit Size:
Pack of 2
Pack of 6
Product Code. 11751243
Brand: WTW™ 208817
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A complete respirometric measuring system for BOD self-check with undiluted samples featuring ranges of up to 4,000mg/L BOD.
  • Easy handling without dilution series
  • Data security by built-in measured value memory
  • Suitable for immediate use
  • Based on pressure management in a closed system - the microorganisms in the sample consume the oxygen and generate CO&sub2; in the process
  • OxiTop™ heads have an AutoTemp function
  • Automatic saving of 5 measured values, manual reading of values after 5 days have elapsed
TRUSTED_SUSTAINABILITY
Specifications
Type BOD Meter, 6 Heads
Display Type LED
Range 0 to 4000 mg/L BSB
For Use With (Application) Respirometric self-check measurement
Includes 6 OxiTop measuring heads (3 yellow, 3 green), 6 PF600 sample bottles, 6 GK 600 Rubber sleeves and 6 RST 600 magnetic stirrers