Learn More
WTW™ OxiTop™ IS BOD Self Check Measurement System
Click to view available options
Type:
BOD Meter, 2 Heads
BOD Meter, 6 Heads
Unit Size:
Pack of 2
Pack of 6
Description
- Easy handling without dilution series
- Data security by built-in measured value memory
- Suitable for immediate use
- Based on pressure management in a closed system - the microorganisms in the sample consume the oxygen and generate CO&sub2; in the process
- OxiTop™ heads have an AutoTemp function
- Automatic saving of 5 measured values, manual reading of values after 5 days have elapsed
Spécification
Spécification
| Type | BOD Meter, 2 Heads |
| Display Type | LED |
| Range | 0 to 4000 mg/L BSB |
| For Use With (Application) | Respirometric self-check measurement |
| Includes | 2 OxiTop measuring heads (yellow, green), 2 PF600 sample bottles, 2 GK 600 Rubber sleeves and 2 RST 600 magnetic stirrers |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu