Learn More
Oxcarbazepine-D4 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | Oxcarbazepine-D4 |
| CAS | 1020719-71-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 2H4 H8 N2 O2 |
| Quantity | 2.5 mg |
| Synonym | Oxcarbazepine D4 |
| InChI Formula | InChI=1S/C15H12N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8H,9H2,(H2,16,19)/i1D,3D,5D,7D |
| SMILES | [2H]c1c([2H])c([2H])c2N(C(=O)N)c3ccccc3C(=O)Cc2c1[2H] |
| IUPAC Name | 1,2,3,4-tetradeuterio-6-oxo-5H-benzo[b][1]benzazepine-11-carboxamide |
| Molecular Weight (g/mol) | 256.29 |
| Show More |
Product Title
Vous avez repéré une opportunité d'amélioration ?