Learn More
Oxazole, 98%, BLD Pharm™
Spécification
Spécification
| CAS | 288-42-6 |
| Molecular Formula | C3H3NO |
| MDL Number | MFCD00009751 |
| Quantity | 10 g |
| SMILES | O1C=NC=C1 |
| Molecular Weight (g/mol) | 69.06 |
| Percent Purity | 98% |
| Chemical Name or Material | Oxazole |