Learn More
Oxalic Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Product Code. 10522572 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
6 Ampules
Unit Size:
1 ampoule
6 ampoules
Product Code. 10522572
Brand: Fisher Chemical J/5650C/05
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemical Identifiers
MDL Number 2573
Specifications
pH 1
Melting Point >160°C
Quantity 1 Ampule
Packaging Ampoule
Physical Form Liquid
compliance-icons
Product Identifier
  • Oxalic acid 0.05M solution (0.5M Concentrate)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.