Learn More
Thermo Scientific™ P2-CST Multiple Gradient Gel Caster Comb, 25 teeth
Description
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
- With an external gradient maker, gels can be cast with ease and great reproducibility
- The design and format of this unit will provide years of service in your lab
Specifications
Specifications
| For Use With (Equipment) | Fisherbrand P2-CST System |
| Type | Frosted Blank Glass |
| Teeth Count | Not Applicable |
| Thickness (English) | 3/16 in. |
| Thickness (Metric) Teeth | Not Applicable |
| Length (Metric) | 20 cm |
| Width (Metric) | 20 cm |