Learn More
Thermo Scientific™ P2-CST Multiple Gradient Gel Caster Comb, 25 teeth
Product Code. 11963055
Click to view available options
Type:
Blank Glass
Caster
Frosted Blank Glass
Offset Glass
Teeth Count:
Not Applicable
Thickness (Metric) Teeth:
Not Applicable
Unit Size:
Each
Product Code. 11963055
Brand: Thermo Scientific™ P218G
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fisherbrand P2-CST System
Gel Size 20 cm to 42 cm x 48 cm
Type Offset Glass
Teeth Count Not Applicable
Thickness (English) 0.18 in.
Thickness (Metric) Teeth Not Applicable
Length (Metric) 18 cm
Width (Metric) 20 cm