Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Product Code. p-4529624 Shop All Thermo Scientific Products
Click to view available options
Gel Size:
14 x 14 cm
24 x 24 cm
Unit Size:
Each
Product Code. 12454637
Brand: Thermo Scientific™ GDF10
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
  • Available in minigel and large format sizes
  • Convenient method of preserving gels by air drying them between two sheets of cellophane
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
Complete kits include two sets of gel-drying frames, 50 sheets of precut cellophane, two solid back plates, and eight side clips; frames and cellphane are also available separately
TRUSTED_SUSTAINABILITY
Specifications
Gel Size 14 x 14 cm
Product Type Gel Drying Kit
Quantity Each
Length (Metric) 14 cm
Width (Metric) 14 cm