Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 System Replacement Parts
Product Code. 11932965
Click to view available options
Type:
External Caster
Gasket
Gel Tray
Includes:
12 Slots, 12 Rapid Load Combs
Gasket Pair
Gel Tray Only
Trays and Combs Not Included
Unit Size:
2 sets
Each
Product Code. 11932965
Brand: Thermo Scientific™ B2CST
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Replace your UVT gel tray, gaskets and external casters with B2 and B3 system replacement parts.
Replace your UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 System Replacement Parts, for use with EasyCast™ Mini Gel Systems.
For use with Owl EasyCast B2 and B3 Gel Systems.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System
Gel Size Mini
Product Line Owl, EasyCast
Type External Caster
Includes Trays and Combs Not Included
Length (Metric) 14 cm
Width (Metric) 12 cm