Learn More
Chemsplash Overshoes (Pairs) PB 6B Elasticated Ankle
GREENER_CHOICE
Product Code. p-200105899
Click to view available options
Size:
Large
X-Large
Unit Size:
200 pairs
Product Code. 30058391
Brand: Chemsplash 2560.400.006
Only null left
Add to basket
Add to basket
Elasticated Overshoe is made from Chemsplash soft and microporous fabric. It is strong and resistant to chemical splash and dust.
Chemsplash Elasticated Overshoe
  • Chemsplash 55GSM Type PB 6B Material
  • 16 in. in Length
  • Silicone and Latex Free
  • Non-Linting
  • Anti-Static Material
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Elasticated Overshoe is made from Chemsplash soft and microporous fabric. It is strong and resistant to chemical splash and dust. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Size Large
Material Microporous Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Seamless
Resistance Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)