Learn More
BINDER™ Overheating Alarm Function for Drying and Heating Chambers
Product Code. 16454169 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 23
BINDER Chambers FD 23
BINDER Chambers FED 400
Unit Size:
Each
Product Code. 16454169
Brand: BINDER™ 80121301
Only null left
Add to basket
Add to basket
The Overheating Alarm Function for Drying and Heating Chambers features a switchable acoustic alarm, with adjustable limit value on the independent temperature safety device.
  • Audible alarm function for overheating
  • Features switchable acoustic alarm
  • Features adjustable limit value
  • Designed for use with BINDER™ Drying and Heating Chambers
Specifications
Description Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
For Use With (Equipment) BINDER Chambers FED 400
Type Adjustable Overheating Alarm
Voltage 400 V