Learn More
R&D Systems™ Ovary Tissue Slides (Endometrioid Adenocarcinoma)- Paraffin
Product Code. 18757383 Shop All Bio Techne Products
Change view
Click to view available options
Species:
Human
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species Unit Size
18757383 Human Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18757383 Supplier R&D Systems™ Supplier No. NBP230292
  • ONLINE EXCLUSIVE PRICE
    255.52€
    213.15€ / Pack of 5
    Save 42.37€
    16% Off
 Restricted Item
Estimated Shipment: 15-12-2026
A Novus product, part of R&D Systems.
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
TRUSTED_SUSTAINABILITY
Specifications
Cross Reactivity Human
Type Ovary Tissue Slides (Endometrioid Adenocarcinoma)
Regulatory Status RUO
Species Human
Tissue Condition Endometrioid Adenocarcinoma
Product Line Novus