Learn More
Cowie Technology™ Oval PTFE Turbo Stir Bars with Rare Earth Core
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
35 mm
40 mm
50 mm
64 mm
Diameter (Metric):
16 mm
20 mm
Unit Size:
Each
Pack of 5
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Samarium-Cobalt magnet core |
| Dimensions (L x W x H) | 50 x 20 x 20 mm |
| For Use With (Equipment) | Round Bottom Flasks |
| Height (Metric) | 20 mm |
| Coating | PTFE |
| Diameter (Metric) | 20 mm |
| Length (Metric) | 50 mm |
| Show More |