missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSCAR Mouse anti-Human, Clone: 259501, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB2004-SP
This item is not returnable.
View return policy
Description
OSCAR Monoclonal specifically detects OSCAR in Human samples. It is validated for Western Blot.Specifications
| OSCAR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8IYS5 | |
| OSCAR | |
| Mouse myeloma cell line NS0-derived recombinant human OSCAR isoform 2 Asp19-Asn229 (Asn158Ser) Accession # Q8IYS5 | |
| 25 μg | |
| Primary | |
| Detects human OSCAR in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse OSCAR. | |
| Human | |
| Purified |
| Western Blot | |
| 259501 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 126014 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction