Learn More
Hanna Instruments™ ORP/pH/Temperature Tester
Product Code. 10305534
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
Product Code. 10305534
Brand: Hanna Instruments™ HI98121
Only null left
Add to basket
Add to basket
Designed to float making it ideal for taking readings virtually anywhere. Hanna Instruments™ ORP/pH/Temperature Tester is a waterproof pH, ORP and temperature meter with the housing completely sealed against humidity. Clears clogs easily with cloth junction.
Waterproof pH, ORP and temperature meter with housing completely sealed against humidity and is designed to float. Replace electrodes with the stainless steel round connector that pulls out 3mm to clear the clog.
  • Monitor both pH and ORP at the same time.
  • Waterproof
  • Automatic Temperature Compensation
  • HOLD feature
  • Simple to use Economical
Specifications
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±0.2
Accuracy (pH) ±0.05
Calibration Automatic
Range (pH) -2 to 16
Resolution (pH) 0.01
Range (mV) ±1000
Range (ORP) ±1000mV
Resolution (mV) 0.1mV
Calibration Points 1 or 2 with 2 sets of standard buffers (pH 4.01, 7.01, 10.01 or pH 4.01, 6.86, 9.18)
Temperature Range (Metric) -5.0°C to +60.0°C
Resolution (Temperature) 0.1°C
Dimensions (L x W x H) 163 x 40 x 26 mm
Weight (Metric) 100 g
Detection Range -2.00 to 16.00 pH
Temperature Compensation Automatic for pH readings
Battery Type 1.5V
Type ORP/pH/Temperature Tester
Battery Life 250 hr.