missing translation for 'onlineSavingsMsg'
Learn More

Ornidazole, TRC

Product Code. 30423644
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30423644

Brand: Toronto Research Chemicals TRCO68850050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ornidazole
CAS 16773-42-5
Purity Grade Notes HPLC
Molecular Formula C7 H10 Cl N3 O3
Quantity 50 mg
Synonym Ornidazole, 1H-Imidazole-1-ethanol','α-(chloromethyl)-2-methyl-5-nitro-, Imidazole-1-ethanol','α-(chloromethyl)-2-methyl-5-nitro- (8CI), α-(Chloromethyl)-2-methyl-5-nitro-1H-imidazole-1-ethanol, (±)-Ornidazole, (±)-Tiberal, 1-(3-Chloro-2-hydroxypropyl)-2-methyl-5-nitroimidazole, 1-Chloro-3-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol, 1-Chloro-3-(2-methyl-5-nitro-imidazol-1-yl)-propan-2-ol, 1-Chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol, Dazolic, Giro, Madelen, NSC 95075, Ornidal, Ornidazole, Ornidone, Ornilox, Ornisid, Ornitop, Ro 7-0207, Tiberal, α-(Chloromethyl)-2-methyl-5-nitro-1-imidazoleethanol
InChI Formula InChI=1S/C7H10ClN3O3/c1-5-9-3-7(11(13)14)10(5)4-6(12)2-8/h3,6,12H,2,4H2,1H3
SMILES Cc1ncc([N+](=O)[O-])n1CC(O)CCl
IUPAC Name 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
Molecular Weight (g/mol) 219.63
Formula Weight 219.0411
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.