Learn More
Ornidazole, 99%, Thermo Scientific™
Interferes with electron transport in metabolic pathways
62.70€ - 220.00€
Chemical Identifiers
| CAS | 16773-42-5 |
|---|---|
| Molecular Formula | C7H10ClN3O3 |
| Molecular Weight (g/mol) | 219.625 |
| MDL Number | MFCD00057960 |
| InChI Key | IPWKIXLWTCNBKN-UHFFFAOYSA-N |
| Synonym | ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole |
| PubChem CID | 28061 |
| ChEBI | CHEBI:75176 |
| IUPAC Name | 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol |
| SMILES | CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15425369
|
Thermo Scientific Alfa Aesar
J63100.06 |
5g |
62.70€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15435369
|
Thermo Scientific Alfa Aesar
J63100.14 |
25g |
220.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in ethanol at 50mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| 16773-42-5 | |
| 219.625 | |
| IPWKIXLWTCNBKN-UHFFFAOYSA-N | |
| 28061 | |
| 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol |
| C7H10ClN3O3 | |
| MFCD00057960 | |
| ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole | |
| CHEBI:75176 | |
| CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-] |
Specifications
| 16773-42-5 | |
| MFCD00057960 | |
| ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole | |
| IPWKIXLWTCNBKN-UHFFFAOYSA-N | |
| 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol | |
| 28061 | |
| 219.63 | |
| Ornidazole |
| C7H10ClN3O3 | |
| 14,6872 | |
| Soluble in ethanol at 50mg/ml | |
| CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-] | |
| 219.625 | |
| CHEBI:75176 | |
| 99% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 240-826-0
missing translation for 'rtecsNumber' : NI5460000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.