Learn More
Thermo Scientific™ Orion™ 2-Cell Conductivity Electrodes
Product Code. 10727564
Change view
Click to view available options
Cell Constant:
0.1 cm^-1
1 cm^-1
10 cm^-1
Measurement Ranges:
0.01 to 300 μS/cm
0.1 μS/cm to 100 mS/cm
0.1 to 100 μS/cm
1 μS/cm to 20 mS/cm
1 μS/cm to 200 mS/cm
10 μS/cm to 200 mS/cm
10 μS/cm to 2000 mS/cm
Connector Types:
8-pin DIN
8-pin Mini-DIN
8-pin Waterproof DIN
Mini-DIN
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cell Constant Measurement Ranges Connector Types unitSize
10727564 0.1 cm^-1 0.1 to 100 μS/cm 8-pin DIN Each
11748196 1 cm^-1 1 μS/cm to 200 mS/cm 8-pin DIN Each
10153012 1 cm^-1 1 μS/cm to 200 mS/cm 8-pin Waterproof DIN Each
11768196 0.1 cm^-1 0.1 μS/cm to 100 mS/cm 8-pin DIN Each
10256152 1 cm^-1 1 μS/cm to 20 mS/cm 8-pin DIN Each
15937431 1 cm^-1 1 μS/cm to 20 mS/cm 8-pin DIN Each
10511833 1 cm^-1 1 μS/cm to 20 mS/cm Mini-DIN Each
10770085 1 cm^-1 10 μS/cm to 200 mS/cm 8-pin Mini-DIN Each
11996079 0.1 cm^-1 0.01 to 300 μS/cm Mini-DIN Each
10113042 10 cm^-1 10 μS/cm to 2000 mS/cm Mini-DIN Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10727564 Supplier Thermo Scientific™ Supplier No. 011020A
Achieve accurate measurements from ultrapure water, sea water, and more with these versatile 2-cell conductivity electrodes, available in a variety of materials and measurement ranges.
The Thermo Scientific™ Orion™ Conductivity electrode 013016MD complies with SM 2510B; D1125-95, -99 (A) for Drinking water measurements and USP<645> Measuring the Conductivity of Pure Water.
 The ThermoFisher Orion Conductivity electrode 013016MD complies with SM 2510B; D1125-95, -99 (A) for Drinking water measurements USP<645> Measuring the Conductivity of Pure Water.
Features
These probes use two electrode cells to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field.
013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly.018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases.011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
Solutions Required
Standards
  • 948206 - 0.1 M lead standard, 475 mL bottle
  • 948207 – 0.1 M sodium sulfate standard for sulfate analysis with the lead ISE 475 mL bottle
Electrode Fill Solution
  • 900062 – Optimum results B electrode filling solution, 5 x 60 mL bottles
Items Included
  • 900062 – Optimum results B electrode filling solution, 1 x 60 mL bottle
  • 948201 – Polishing strip, 2 count
Warranty Period
  • 12 months from date of purchase
TRUSTED_SUSTAINABILITY
Specifications
Cell Constant 0.1 cm^-1
Connector Types 8-pin DIN
For Use With (Equipment) 555A, 550A, 162A, 136A, 135A, 130A Meters
Type 2-Electrode Conductivity Cells
Description Orion 2 Cell Platinized Conductivity Probe
Material Glass, Platinum
Measurement Ranges 0.1 to 100 μS/cm