Learn More
- Home
- Products
- pH and Electrochemistry
- Benchtop Titrators
- Thermo Scientific™ Orion Star T900 Series Titrator Accessories
Thermo Scientific™ Orion Star T900 Series Titrator Accessories
Product Code. 15872107| Artikelnummer. | Type | unitSize |
|---|---|---|
| 15872107 | Burette Cover | Each |
| 15882107 | 10 mL Burette | Each |
| 15892107 | 20 mL Burette | Each |
| 15802117 | 50 mL Burette | Each |
| 15812117 | Reagent Bottle Holder | Each |
| 15822117 | 1 L Plastic Bottle | Each |
| 15832117 | 1 L Glass Bottle | Each |
| 15842117 | 12 x 1 L Plastic Bottles | Each |
| 15852117 | 12 x 1 L Glass Bottles | Each |
| 15862117 | Reagent Bottle Cap (GL38) | Each |
| 15872117 | Reagent Bottle Cap (GL45) | Each |
| 15882117 | Pint Bottle Cap | Each |
| 15892117 | Dispenser Probe | Each |
| 15802127 | Dispenser Verification Kit | Each |
| 15812127 | Electrode Holder Assembly | Each |
| 15822127 | Stopper | Each |
| 15832127 | Cable Management | Each |
| 15842127 | EU Power Supply | Each |
| 15852127 | UK/Singapore Power Supply | Each |
| 15862127 | Standard Tubing Kit | Each |
| 15872127 | Light-blocking Tubing Kit | Each |
| 15882127 | Drying Tube | Each |
| 15892127 | Replacemet Burette | Each |
| 15802137 | Replacement Reagent Cap | Each |
| 15812137 | Replacement Dispenser Probe | Each |
| 15822137 | User Manual | Each |
| 15832137 | USB Cable | Each |
| 15842137 | Value Replacement | Each |
Beschreibung
Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.
•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable
Spezifikation
Spezifikation
| Description | Orion Star T900 series burette cover |
| Type | Burette Cover |
| For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.