Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Product Code. 11727727
Click to view available options
Type:
Ammonia
Chlorine (free)
Chlorine (total)
Copper (free)
Iron (ferro)
Iron (total)
Manganese
Molybdate/Molybdenum
Nitrite
Phosphate (ortho)
Silica
Sulfate
Range:
0.01 to 0.3 mg/L N
0.01 to 0.8 mg/L N
0.02 to 1.8 mg/L Fe
0.02 to 2 mg/L Cl2
0.02 to 3 mg/L Fe
0.05 to 5 mg/L Cu
0.06 to 2.5 mg/L PO4
0.1 to 18 mg/L Mn
0.5 to 66 mg/L MoO4
1 to 90 mg/L SiO2
5 to 100 mg/L SO4
Unit Size:
100 tests
Each
Pack of 100
Product Code. 11727727
Brand: Thermo Scientific™ AC4P42
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters
TRUSTED_SUSTAINABILITY
Specifications
Type Molybdate/Molybdenum
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Includes Set of 3: 100 Molybdenum HR1 Powder Packs, 100 Molybdenum HR2 Powder Packs, 100 Molybdenum HR3 Powder Packs
Packaging Powder Pack
Quantity 100 tests
Range 0.5 to 66 mg/L MoO4
Recommended Accessories AQUAfast 24 mm round vials (Cat. No. AC2V24)
Test Method Mercaptoacetic Acid
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.