Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Product Code. 11717727
Change view
Click to view available options
Type:
Ammonia
Chlorine (free)
Chlorine (total)
Copper (free)
Iron (ferro)
Iron (total)
Manganese
Molybdate/Molybdenum
Nitrite
Phosphate (ortho)
Silica
Sulfate
Range:
0.01 to 0.3 mg/L N
0.01 to 0.8 mg/L N
0.02 to 1.8 mg/L Fe
0.02 to 2 mg/L Cl2
0.02 to 3 mg/L Fe
0.05 to 5 mg/L Cu
0.06 to 2.5 mg/L PO4
0.1 to 18 mg/L Mn
0.5 to 66 mg/L MoO4
1 to 90 mg/L SiO2
5 to 100 mg/L SO4
Unit Size:
100 tests
Each
Pack of 100
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Range unitSize
11717727 Copper (free) 0.05 to 5 mg/L Cu Each
11707727 Ammonia 0.01 to 0.8 mg/L N Each
15159192 Molybdate/Molybdenum 0.5 to 66 mg/L MoO4 Pack of 100
11727727 Molybdate/Molybdenum 0.5 to 66 mg/L MoO4 Each
11737727 Nitrite 0.01 to 0.3 mg/L N Each
11747727 Manganese 0.1 to 18 mg/L Mn Each
11712248 Silica 1 to 90 mg/L SiO2 Pack of 100
11757727 Chlorine (free) 0.02 to 2 mg/L Cl2 Pack of 100
11767727 Chlorine (total) 0.02 to 2 mg/L Cl2 Each
11777727 Iron (ferro) 0.02 to 3 mg/L Fe Each
11787727 Iron (total) 0.02 to 1.8 mg/L Fe Each
11797727 Sulfate 5 to 100 mg/L SO4 100 tests
11707737 Phosphate (ortho) 0.06 to 2.5 mg/L PO4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11717727 Supplier Thermo Scientific™ Supplier No. AC4P29
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters
TRUSTED_SUSTAINABILITY
Specifications
Type Copper (free)
EPA Approved Methods Wastewater: Method 8506
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Includes 100 Cu 1 Powder Packs
Packaging Powder Pack
Quantity 100 tests
Range 0.05 to 5 mg/L Cu
Recommended Accessories AQUAfast 24 mm round vials (Cat. No. AC2V24)
Test Method Bicinchoninate
compliance-icons
Product Identifier
  • AQUAfast Copper 100pcs
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ammonium acetate, sodium bisulphite, ammonium oxalate, acetone, bathocuproine
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.