Learn More
Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder with Star A series portable meter armor and sleeve to protect your pH electrode in the field
Brand:  Thermo Scientific™ STARA-ESPH
Additional Details : Weight : 0.90720kg
Product Code. 11671489
  • 27.40€ / Each
In Stock
Description
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder with the Star A series portable meter armor (Cat. No. STARA-AR) and storage sleeve (Cat. No. 810017) to protect, transport and store your sensors in the field. This pH electrode holder can be attached to either side of the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeve.
Specifications
Specifications
Orion Star A series pH electrode holder for portable meter protective armor
Meter Accessories
pH electrode storage sleeve (Cat. No. 810017)