Learn More
Thermo Scientific™ Orion™ Model 900A Printer Replacement Parts
Replace printer paper and rechargeable battery pack with Thermo Scientific™ Orion™ Model 900A Printer Replacement Parts, for use with the Orion™ 900A thermal printer.
Brand: Thermo Scientific™ 0900A4
103.10 EUR valid until 2023-02-05
Use promo code "19086" to get your promotional price.
Additional Details : Weight : 0.19800kg
Description
Replace printer paper and rechargeable battery packs with Thermo Scientific™ Orion™ Model 900A Printer Replacement Parts, for use with the Orion™ 900A thermal printer.
Specifications
Replacement Paper for Orion 900A Printer |