Learn More
Thermo Scientific™ Orion Lab Star Bench Meter Accessories
Description
Select Orion Lab Star series meter accessories to support your Orion Lab Star PH111 pH/mV meter, Orion Lab Star EC112 conductivity meter or Orion Lab Star DO113 dissolved oxygen meter. Utilize the meter dust cover accessory for extended meter storage. Select a replacement meter-attached electrode holder, universal power adapter or computer cable when needed. The Orion Lab Star series pH meter, conductivity meter and DO meter are ideal for simplified, budget-friendly lab testing and data recording.
Orion Lab Star Series Meter Accessories Features
- LSTAR-ARM Orion Lab Star series meter-attached electrode stand allows you to replace your electrode stand as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
- LSTAR-PWR Orion Lab Star series universal power supply, 100-240V 50/60 Hz, allows you to replace your power adapter as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
- LSTAR-USB Orion Lab Star series USB computer cable allows you to replace your computer cable as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
- LSTAR-CVR Orion Lab Star series meter dust cover protects your meter from dust and debris while in storage and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
Additional Accessories Compatible with Orion Lab Star Series Meters
- STARA-106 ink-ribbon compact printer, 100-240V 50/60 Hz
Spécification
Spécification
| Description | Orion Lab Star series meter dust cover |
| Type | Dust Cover |
| Certifications/Compliance | None |
| Compatibility | Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.