Learn More
Thermo Scientific™ Orion™ Ion Selective Electrode Accessories
Product Code. 10461213
Change view
Click to view available options
For Use With (Equipment):
ISE Half Cell Electrodes
Ion Selective Electrode
Type:
Handle
Polishing Strips
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Unit Size
10461213 ISE Half Cell Electrodes Handle Each
10428722 Ion Selective Electrode Polishing Strips Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10461213 Supplier Thermo Scientific™ Supplier No. 9300BNWP
  • ONLINE EXCLUSIVE PRICE
    625.00€
    581.00€ / Pack of 1
    Save 44.00€
    7% Off
Estimated Shipment: 05-11-2026
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Electrodes handles are used with plastic membrane combination or half-cell electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Description Handle for ISE Half Cell Plastic Membrane Electrodes wth BNC waterproof connector
For Use With (Equipment) ISE Half Cell Electrodes
Type Handle