Learn More
Thermo Scientific™ Orion™ Iodide Ion Selective Electrodes
Product Code. 10288420
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw cap
Unit Size:
Each
Product Code. 10288420
Brand: Thermo Scientific™ 9453SC
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Get fast, stable and reliable iodide ions readings in aqueous solutions.
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
  • Calibration standards catalog number: 945306 - 0.1M NaI Required ISA catalog number: 940011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Screw cap
Description With screw cap connector
For Use With (Equipment) ISE Meter
Type Combination ISE