Learn More
Thermo Scientific™ Orion™ Iodide Ion Selective Electrodes
missing translation for 'orderingAttributeHoverText'
Connector Types:
BNC
BNC (Waterproof)
Screw cap
missing translation for 'unitSize'
Each
Description
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
- Calibration standards catalog number: 945306 - 0.1M NaI Required ISA catalog number: 940011
Specifications
Specifications
| Connector Types | BNC |
| Description | With BNC connector |
| For Use With (Equipment) | ISE Meter |
| Type | Solid state half-cell |