Learn More
Thermo Scientific™ Orion™ Gas-Sensing ISE membranes
Product Code. 10408532
Change view
Click to view available options
For Use With (Equipment):
Carbon Dioxide Electrode, 9502BNMP
HP Ammonia Electrode, 9512HPBNWP
HP Ammonia Electrode, 9512HPBNWP, 9512HP
Standard Ammonia Electrode, 9512BNWP, 9512
Unit Size:
Each
Pack of 20
Pack of 3
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10408532 Carbon Dioxide Electrode, 9502BNMP Each
10292441 Standard Ammonia Electrode, 9512BNWP, 9512 Pack of 20
10758971 Standard Ammonia Electrode, 9512BNWP, 9512 Pack of 3
10554454 HP Ammonia Electrode, 9512HPBNWP, 9512HP Pack of 20
10165673 HP Ammonia Electrode, 9512HPBNWP Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10408532 Supplier Thermo Scientific™ Supplier No. 950204
  • ONLINE EXCLUSIVE PRICE
    233.00€
    217.00€ / Pack of 1
    Save 16.00€
    6% Off
View all promotions
In Stock
Add to basket
Add to basket
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
TRUSTED_SUSTAINABILITY
Specifications
Description For Carbon Dioxide Electrode 9502BNMP; Four membranes with O-rings
For Use With (Equipment) Carbon Dioxide Electrode, 9502BNMP
Type Gas-Sensing ISE Membrane