Learn More
Thermo Scientific™ Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Product Code. 15169192
missing translation for 'orderingAttributeHoverText'
Type:
Ammonia
COD
COD High Range
COD Low Range
COD Mid Range
Fluoride
Manganese
Nitrate
Nitrogen, Total
Phosphate
pH
Range:
0 to 150 mg/L O2
0 to 1500 mg/L O2
0 to 15000 mg/L O2
0.01 to 0.7 mg/L Mn
0.02 to 1.1 mg/L P
0.02 to 1.6 mg/L P
0.02 to 2.5 mg/L N
0.05 to 2 mg/L F
0.06 to 5 mg/L PO4
0.5 to 25 mg/L N
1 to 30 mg/L N
1 to 50 mg/L N
5 to 150 mg/L N
6.5 to 8.4 pH
missing translation for 'unitSize'
25 tests
Each
Pack of 100
Pack of 2
Pack of 25
Pack of 50
Código de producto. 15169192
Marca: Thermo Scientific™ AC4P54
Este artículo no se puede devolver. Vea la política de devoluciones
Perform colorimetric testing including fluoride, COD, total nitrogen and total phosphate using the Thermo Scientific™ Orion™ AQUAfast Liquid Reagents, Digestion and Reaction Tubes.
Use Thermo Scientific™ Orion™ AQUAfast Digestion Tubes, Reaction Tubes and Liquid Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass and fit the Orion™ AQUAfast COD165 thermoreactor. Digestion tubes measure COD, total nitrogen and total phosphate. Reaction tubes measure ammonia, nitrate and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine and pH.
Changes in nutrient levels can alter the delicate balance in ecosystems and contaminate drinking water. Tube tests make it easy to determine concentrations of species in samples.
Digestion tube tests and reaction tube tests are preprogrammed in the Orion AQ3700 meter. Some of the listed tests have only liquid components, like our AC2009 SPADNS test for fluoride, and some have liquid and powder or liquid and tablet reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Description AQUAfast manganese LR reagent powder and liquid set
Format Powder + Liquid
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter, Orion AQUAfast
Includes Set of 3: 100 Ascorbic Acid Powder Packs, Alkaline-Cyanide Solution, PAN Indicator Solution
Quantity 50 Tests
Range 0.01 to 0.7 mg/L Mn
Recommended Accessories AQUAfast 24mm round vials (Cat. No. AC2V24)
Test Method PAN
Type Manganese
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado