Learn More
Orientin, TRC
Product Code. 30133475
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133475 50 mg 50mg
30010856 25 mg 25mg
30009261 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133475 Supplier Toronto Research Chemicals Supplier No. TRCO67490050MG
 Restricted Item
Estimated Shipment: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28608-75-5
Molecular Formula C21 H20 O11
Molecular Weight (g/mol) 448.38
Synonym Orientin (6CI,7CI,8CI), 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-8-β-D-glucopyranosyl-5,7-dihydroxy-, 2-(3,4-Dihydroxyphenyl)-8-β-D-glucopyranosyl-5,7-dihydroxy-4H-1-benzopyran-4-one, 8-β-D-Glucopyranosyl-3',4',5,7-tetrahydroxyflavone, Luteolin 8-C-glucoside, Luteolin 8-C-β-D-glucopyranoside, Luteolin 8-C-β-glucopyranoside, Luteolin-8-C-glucoside, Luteolin-8-glucoside, Lutexin, Orientin (flavone)
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]chromen-4-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc(O)c3C(=O)C=C(Oc23)c4ccc(O)c(O)c4
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C21H20O11/c22-6-14-17(28)18(29)19(30)21(32-14)16-11(26)4-10(25)15-12(27)5-13(31-20(15)16)7-1-2-8(23)9(24)3-7/h1-5,14,17-19,21-26,28-30H,6H2/t14-,17-,18+,19-,21+/m1/s1
Formula Weight 448.1006
Percent Purity >95%
Chemical Name or Material Orientin
Grade HPLC