Learn More
3M™ Orange SMS Based Fabric Category III Protective Coveralls
Product Code. p-8003857
Click to view available options
Size:
Small
Large
Unit Size:
Pack of 20
Product Code. 11933754
Brand: 3M™ 7000089634
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel
Seam Type Serged
Sterility Non-sterile
Protection Class Category III
Height (Metric) 174 to 181 cm