missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Orange Serum Nutrient Pad and Gridded MF, pH 5.5

Product Code. 15902510 Shop All Sartorius Products
Click to view available options
:
0.45 μm pore size discs
This item is not returnable. View return policy

Product Code. 15902510

Brand: Sartorius Stedim Biotech™ 1409450RDN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dehydrated media pads in petri dishes , pre-sterilized, along with suitable membrane filters for economical, time-saving microbiological quality control.

  • Nutrient pads with gridded membranes
  • Trouble-free storage up to 24 months
  • Compliance with international standards and references
  • Validation guide available
  • Highly versatile - multiple applications
  • Economical by time- and cost-savings
  • Easy handling
  • Membranes suitable for use with e.motion dispenser
  • For the detection and count of acid-tolerant microorganisms in beverages, water and raw materials
TRUSTED_SUSTAINABILITY

Specifications

Color White
Format 0.45μm Pore Size Disc
Media Type Orange Serum (pH 5.5)
Sterility Sterile
Target Organism Class Acid-tolerant Microorganisms
Grid Color Black
Packaging Type Individually wrapped MF for use with dispenser, 10 x 10/bag dishes with NP
Pore Size 0.45μm
Quantity 100/Pk.
Diameter (Metric) 50 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.