Learn More
Macherey-Nagel™ Optima™ 5 Performance GC Capillary Column
Product Code. 10194701
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Film Thickness:
0.1 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
3 μm
5 μm
Unit Size:
Each
Pack of 15
Pack of 50
Product Code. 10194701
Brand: Macherey-Nagel™ 726934.15
Only null left
Specifications
Diameter (Metric) 0.5 mm
Type High Performance GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Optima™ 5
USP Type G27
For Use With (Application) Gas Chromatography (GC)
Temperature Range 340°C
Film Thickness 5 μm
Length (Metric) 15 m
Quantity 1 Each